missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Di-2-pyridyl Carbonate 98.0+%, TCI America™
$68.80 - $200.14
Chemical Identifiers
CAS | 1659-31-0 |
---|---|
Molecular Formula | C11H8N2O3 |
Molecular Weight (g/mol) | 216.196 |
MDL Number | MFCD00191407 |
InChI Key | GCSAXWHQFYOIFE-UHFFFAOYSA-N |
Synonym | Carbonic Acid Di-2-pyridyl Ester |
PubChem CID | 2757370 |
IUPAC Name | dipyridin-2-yl carbonate |
SMILES | C1=CC=NC(=C1)OC(=O)OC2=CC=CC=N2 |
Chemical Identifiers
1659-31-0 | |
216.196 | |
GCSAXWHQFYOIFE-UHFFFAOYSA-N | |
2757370 | |
C1=CC=NC(=C1)OC(=O)OC2=CC=CC=N2 |
C11H8N2O3 | |
MFCD00191407 | |
Carbonic Acid Di-2-pyridyl Ester | |
dipyridin-2-yl carbonate |
Specifications
White-Yellow | |
1g | |
C11H8N2O3 | |
Carbonic Acid Di-2-pyridyl Ester | |
C1=CC=NC(=C1)OC(=O)OC2=CC=CC=N2 | |
216.196 | |
216.20 | |
≥98.0% (T) |
90°C | |
1659-31-0 | |
MFCD00191407 | |
GCSAXWHQFYOIFE-UHFFFAOYSA-N | |
dipyridin-2-yl carbonate | |
2757370 | |
Crystal-Powder at 20°C | |
Di-2-pyridyl Carbonate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator