Please login to your online account to display your discounted pricing

Di-2-ethylhexylamine, 99%, ACROS Organics™

$72.36 - $72.36

Chemical Identifiers

Chemical Name or Material Di-2-ethylhexylamine
Name Note 99%
CAS 106-20-7
Assay 98.5 to 101.5% (Total Base), ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C16H35N
Linear Formula (CH3(CH2)3CH(C2H5)CH2)2NH
Formula Weight 241.46
MDL Number MFCD00009489
Synonym Bis(2-ethylhexyl)amine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC271960010

  SDS
thermo fisher scientific
271960010
Glass bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC271962500

  SDS
thermo fisher scientific
271962500
Glass bottle 250mL Each for $72.36

AC271960025

  SDS
thermo fisher scientific
AC271960025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay 98.5 to 101.5% (Total Base), ≥98.5% (GC)
Physical Form Liquid
Formula Weight 241.46
Quantity 1L
Color Clear colorless to slightly yellow
EINECS Number 203-372-4
TSCA TSCA
Density 0.8000g/mL
RTECS Number IH6825000
Packaging Glass bottle
Refractive Index 1.4415 to 1.4435 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Di-2-ethylhexylamine
Name Note 99%
Assay Percent Range 99%
Molecular Formula C16H35N
Linear Formula (CH3(CH2)3CH(C2H5)CH2)2NH
MDL Number MFCD00009489
CAS 106-20-7
Synonym Bis(2-ethylhexyl)amine
Infrared Spectrum Authentic