Learn More
Di-2-ethylhexylamine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC271960025
Specifications
Di-2-ethylhexylamine | |
128°C | |
Glass bottle | |
0.8 | |
281.0°C | |
-60.0°C | |
99% | |
98.5% min. (GC) | |
(CH3(CH2)3CH(C2H5)CH2)2NH | |
04,388 | |
Solubility in water: <0.2g/L (20 c) in water. Other solubilities: soluble in hydrocarbons | |
CCCCC(CC)CNCC(CC)CCCC | |
241.46 | |
241.46 | |
99% |
0.8000g/mL | |
Authentic | |
1.4415 to 1.4435 | |
3.7 mPa.s (20°C) | |
Colorless to Yellow | |
2.5L | |
106-20-7 | |
C16H35N | |
MFCD00009489 | |
1-hexanamine, 2-ethyl-n-2-ethylhexyl, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, bis 2-ethylhexyl amine, bis-2-ethylhexylamine, ccris 4619, di 2-ethylhexyl amine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, diisooctylamine | |
SAIKULLUBZKPDA-UHFFFAOYSA-N | |
2-ethyl-N-(2-ethylhexyl)hexan-1-amine | |
7791 | |
Liquid |
Chemical Identifiers
106-20-7 | |
241.46 | |
SAIKULLUBZKPDA-UHFFFAOYSA-N | |
7791 | |
CCCCC(CC)CNCC(CC)CCCC |
C16H35N | |
MFCD00009489 | |
1-hexanamine, 2-ethyl-n-2-ethylhexyl, 2,2'-diethyldihexylamine, 2,2'-diethylhexylamine, bis 2-ethylhexyl amine, bis-2-ethylhexylamine, ccris 4619, di 2-ethylhexyl amine, di-2-ethylhexyl amine, dihexylamine, 2,2'-diethyl, diisooctylamine | |
2-ethyl-N-(2-ethylhexyl)hexan-1-amine |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-372-4
RTECSNumber : IH6825000
TSCA : TSCA