missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DHB MALDI Matrix 2,5-Dihydroxybenzoic acid, ProteoSpec™ Ricca Chemical

MALDI Matrices

Manufacturer:  Ricca Chemical Company RIRMB1510224X1CM

 View more versions of this product

Catalog No. MB15102241C



Specifications

Specifications

ProteoSpec™ DHB MALDI Matrix 2,5-Dihydroxybenzoic acid
White to Yellow
24 x 1mg
100
C7H6O4
Soluble in water
C1=CC(=C(C=C1O)C(=O)O)O
154.121
CHEBI:17189
Solid
Molecular Biology
Glass Ampule
199°C to 200°C
490-79-9
100
gentisic acid, hydroquinonecarboxylic acid, benzoic acid, 2,5-dihydroxy, 5-hydroxysalicylic acid, gensigen, gensigon, gentisate, 2,5-dioxybenzoic acid, 2,5-dhba, gentisinic acid
WXTMDXOMEHJXQO-UHFFFAOYSA-N
2,5-dihydroxybenzoic acid
3469
154.12
99.0% min.

Chemical Identifiers

490-79-9
154.121
gentisic acid, hydroquinonecarboxylic acid, benzoic acid, 2,5-dihydroxy, 5-hydroxysalicylic acid, gensigen, gensigon, gentisate, 2,5-dioxybenzoic acid, 2,5-dhba, gentisinic acid
CHEBI:17189
C1=CC(=C(C=C1O)C(=O)O)O
C7H6O4
WXTMDXOMEHJXQO-UHFFFAOYSA-N
3469
2,5-dihydroxybenzoic acid
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Refridgerated

Documents