Please login to your online account to display your discounted pricing
GE Healthcare Dharmacon DharmaFECT™-2 siRNA Transfection Reagent
Most broadly applicable DharmaFECT formulation for optimal siRNA or microRNA transfection into a wide range of cell types for successful RNAi experiments
$186.56 - $898.09
Specifications
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
View More Specs
Products
Catalog Number Mfr. No. Product Type Quantity Price Quantity    
T200201
ge healthcare dharmacon
T-2002-01
DharmaFECT-2 Reagent 0.2mL Each for $186.56
T200203
ge healthcare dharmacon
T-2002-03
DharmaFECT-2 Reagent 1.5mL Each for $898.09
Description & Specifications
Specifications
For Use With (Application) High efficiency of siRNA and microRNA delivery with minimal impact on cell viability across a broad range of cell types for successful RNAi experiments
  • Optimized for small RNA (siRNA or microRNA) delivery
  • DharmaFECT provides four specialized lipid-based formulations for successful delivery to more cell lines than any other single reagent
  • DharmaFECT 1 is the most all-purpose transfection reagent, demonstrating efficient, low-toxicity delivery to over 80% of validated cell types
  • DharmaFECT 2, 3, and 4 offer distinct formulations to support a wider range of cell types, and permit more thorough optimization of transfection for high-value experiments and screening projects