Please login to your online account to display your discounted pricing

Tocris™ DH 97


MT2 receptor antagonist

Manufacturer: tocris™  121810

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 121810

  • / Each
 

Description & Specifications

Specifications

Inhibitors DH 97
Molecular Weight 334.46
Quantity 10mg
Product Type DH 97

A relatively potent MT2 melatonin receptor antagonist (pKi value = 8.03), displaying 89- and 229-fold selectivity over MT1 and GPR50 (melatonin-related orphan receptor) respectively.

  • Chemical Name: N-Pentanoyl-2-benzyltryptamine
  • Formula: C22H26N2O
  • CAS Number: 220339-00-4
  • Solubility: Soluble to 100mM in DMSO