missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ DH 97

MT2 receptor antagonist

$181.19 - $734.17

Specifications

Product Type DH 97
Inhibitors DH 97
Molecular Weight 334.46
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
121810 Tocris™
121810
10mg Each for $181.19
121850 Tocris™
121850
50mg Each for $734.17
Specifications

Description

A relatively potent MT2 melatonin receptor antagonist (pKi value = 8.03), displaying 89- and 229-fold selectivity over MT1 and GPR50 (melatonin-related orphan receptor) respectively.
  • Chemical Name: N-Pentanoyl-2-benzyltryptamine
  • Formula: C22H26N2O
  • CAS Number: 220339-00-4
  • Solubility: Soluble to 100mM in DMSO
Description & Specifications

Specifications

DH 97
DH 97
334.46