Please login to your online account to display your discounted pricing

DFMO, Tocris

Ornithine decarboxylase inhibitor

$88.05 - $361.37

Chemical Identifiers

Chemical Name or Material DFMO
Assay Percent Range >98%
Molecular Formula C6H12F2N2O2·HCl
Formula Weight 218.63
Synonym DL-2-(Difluoromethyl)-ornithine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

276110

 
r & d systems
RDS276110
10mg Each for $88.05

276150

 
r & d systems
RDS276150
50mg Each for $361.37
Description & Specifications

Specifications

Formula Weight 218.63
Quantity 10mg
CAS 68278-23-9
Chemical Name or Material DFMO
Assay Percent Range >98%
Molecular Formula C6H12F2N2O2·HCl
Recommended Storage Desiccate at Room Temperature
Synonym DL-2-(Difluoromethyl)-ornithine hydrochloride
Safety and Handling

Desiccate at Room Temperature