Please login to your online account to display your discounted pricing

DFB, Tocris

Allosteric potentiator at mGlu5

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material DFB
Assay Percent Range >99%
Molecular Formula C14H10F2N2
Formula Weight 244.24
Synonym [(3-Fluorophenyl)methylene]hydrazone-3-fluorobenzaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

162510

 
r & d systems
RDS162510
10mg Each for $121.23

162550

 
r & d systems
RDS162550
50mg Each for $509.13
Description & Specifications

Specifications

Formula Weight 244.24
Quantity 10mg
Molecular Formula C14H10F2N2
Synonym [(3-Fluorophenyl)methylene]hydrazone-3-fluorobenzaldehyde
Recommended Storage Store at +4°C
Chemical Name or Material DFB
Assay Percent Range >99%
CAS 15332-10-2
Safety and Handling

Store at +4°C