Please login to your online account to display your discounted pricing

Dextrose U.S.P. Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Dextrose
Name Note Anhydrous
CAS 50-99-7
Purity Grade USP
Physical Form Granular
Molecular Formula C6H12O6
Formula Weight 180.16g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK4908500

 
avantor performance
490804
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Dextrose
Name Note Anhydrous
CAS 50-99-7
Purity Grade USP
Physical Form Granular
Molecular Formula C6H12O6
Formula Weight 180.16g/mol