Please login to your online account to display your discounted pricing

Dextrose U.S.P. Granular, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  490804

 View more versions of this product

Catalog No. MK4908500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Dextrose
Name Note Anhydrous
CAS 50-99-7
Purity Grade USP
Physical Form Granular
Molecular Formula C6H12O6
Formula Weight 180.16g/mol

Specifications

Chemical Name or Material Dextrose
Name Note Anhydrous
CAS 50-99-7
Purity Grade USP
Physical Form Granular
Molecular Formula C6H12O6
Formula Weight 180.16g/mol