missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dextrose U.S.P. GenAR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 50-99-7
Physical Form Granular
Molecular Formula C6H12O6
Formula Weight 180.16
Chemical Name or Material Dextrose, Anhydrous
Grade USP
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7730212 Avantor Performance
773006
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

50-99-7
C6H12O6
Dextrose, Anhydrous
Granular
180.16
USP

Specifications

180.16
C6H12O6
50-99-7
Dextrose, Anhydrous
Granular
Poly
2.5kg
USP