missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dextrose Monohydrate (Powder/USP), Fisher Chemical™
$1418.93 - $4350.57
Chemical Identifiers
CAS | 77938-63-7 |
---|---|
Molecular Formula | C6H14O7 |
Molecular Weight (g/mol) | 198.17 |
MDL Number | MFCD00149450 |
InChI Key | SPFMQWBKVUQXJV-UHFFFAOYNA-N |
Synonym | d-glucose monohydrate, dextrose monohydrate, d-glucose, monohydrate, glucose monohydrate, d-+-glucose monohydrate, d +-glucose monohydrate, unii-lx22yl083g, dextrosum glucosum monohydricum, glucose water, dextrose hydrate |
PubChem CID | 22814120 |
IUPAC Name | 2,3,4,5,6-pentahydroxyhexanal hydrate |
SMILES | O.OCC(O)C(O)C(O)C(O)C=O |
Chemical Identifiers
77938-63-7 | |
198.17 | |
SPFMQWBKVUQXJV-UHFFFAOYNA-N | |
22814120 | |
O.OCC(O)C(O)C(O)C(O)C=O |
C6H14O7 | |
MFCD00149450 | |
d-glucose monohydrate, dextrose monohydrate, d-glucose, monohydrate, glucose monohydrate, d-+-glucose monohydrate, d +-glucose monohydrate, unii-lx22yl083g, dextrosum glucosum monohydricum, glucose water, dextrose hydrate | |
2,3,4,5,6-pentahydroxyhexanal hydrate |
Specifications
77938-63-7 | |
White | |
Poly Pail | |
CH2OHCHO(CHOH)3CHOH·H2O | |
3 kg | |
Meets Requirements (USP) | |
O.OCC(O)C(O)C(O)C(O)C=O | |
198.17 | |
198.17 | |
USP | |
Dextrose Monohydrate |
83°C | |
1.54g/mL | |
C6H14O7 | |
MFCD00149450 | |
d-glucose monohydrate, dextrose monohydrate, d-glucose, monohydrate, glucose monohydrate, d-+-glucose monohydrate, d +-glucose monohydrate, unii-lx22yl083g, dextrosum glucosum monohydricum, glucose water, dextrose hydrate | |
SPFMQWBKVUQXJV-UHFFFAOYNA-N | |
2,3,4,5,6-pentahydroxyhexanal hydrate | |
22814120 | |
Negligible | |
Solid |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0