Please login to your online account to display your discounted pricing

Dextrose Monohydrate (Crystalline Powder/USP/EP/BP), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  FLD12-500

 View more versions of this product

Catalog No. D12-500

  • / Each
 

Description & Specifications

Chemical Identifiers

Acidity Pass Test (USP)
Acidity Alkalinity Pass Test (BP)
Additional Information Vapor Pressure: Negligible
Appearance of Solution Pass Test (BP)
Arsenic (As) 1ppm max. (BP)
Barium (Ba) Pass Test (BP)
Calcium (Ca) 200ppm max. (BP)
Chloride 0.018% max. (USP)
Dextrins Pass Test (USP)
Heavy Metals (as Pb) 5ppm max. (USP)
Identification Pass Test (BP)
Ignition Residue 0.1% max. (USP)
Lead (Pb) 0.5ppm max. (EP)
Packaging Glass Bottle
Pyrogens Pass Test (BP)
Residual Solvents Meets Requirements (BP)
Specific Rotation +52.5 to +53.3° (+ or -) (BP)
Soluble Starch and Sulfites Pass Test (USP)
Sulfate 0.025% max. (USP)
Sulfite 15ppm max. (BP)
Sulfated Ash 0.1% max. (BP)
Water 7.0 to 9.5% (BP/EP), 7.5 to 9.5% (USP)
Color White
Melting Point 83°C / 181.4°F
Quantity 500g
Chemical Name or Material Dextrose Monohydrate
CAS 77938-63-7
Purity Grade USP/EP/BP
Physical Form Solid
Molecular Formula C6H12O6 · H2O
Linear Formula CH2OHCHO(CHOH)3CHOH·H2O
Formula Weight 198.17g/mol
MDL Number MFCD00149450
Synonym D-Glucose Monohydrate
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

Specifications

Chemical Name or Material Dextrose Monohydrate
CAS 77938-63-7
Purity Grade USP/EP/BP
Physical Form Solid
Molecular Formula C6H12O6 · H2O
Linear Formula CH2OHCHO(CHOH)3CHOH·H2O
Formula Weight 198.17g/mol
MDL Number MFCD00149450
Synonym D-Glucose Monohydrate
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents