Please login to your online account to display your discounted pricing

Dextrose Monohydrate (Crystalline Powder/USP/EP/BP), Fisher Chemical


Quantity 50kg
Packaging Fiber Drum

Manufacturer: thermo fisher scientific  FLD12-50

 View more versions of this product

Catalog No. D12-50

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 198.17
Purity Grade BP/EP/USP
Melting Point 83°C
Quantity 50kg
CAS 77938-63-7
Physical Form Solid
Molecular Formula C6H12O6 · H2O
Water 7.0 to 9.5% (BP/EP), 7.5 to 9.5% (USP)
Color White
Sulfate 0.025% max. (USP)
Soluble Starch and Sulfites Pass Test (USP)
Identification Pass Test (BP)
Heavy Metals (as Pb) 5ppm max. (USP)
Calcium (Ca) 200ppm max. (BP)
Sulfated Ash 0.1% max. (BP)
Residual Solvents Meets Requirements (BP)
Pyrogens Pass Test (BP)
Appearance of Solution Pass Test (BP)
Acidity Alkalinity Pass Test (BP)
Specific Rotation +52.5 to +53.3° (+ or -) (BP)
Lead (Pb) 0.5ppm max. (EP)
Chloride 0.018% max. (USP)
Sulfite 15ppm max. (BP)
Dextrins Pass Test (USP)
Arsenic (As) 1ppm max. (BP)
Health Hazard 2 CAUTION!
Chemical Name or Material Dextrose Monohydrate
Linear Formula CH2OHCHO(CHOH)3CHOH·H2O
MDL Number MFCD00149450
Additional Information Vapor Pressure: Negligible
Packaging Fiber Drum
Ignition Residue 0.1% max. (USP)
Barium (Ba) Pass Test (BP)
Acidity Pass Test (USP)
Synonym D-Glucose Monohydrate
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Dextrose Monohydrate
CAS 77938-63-7
Purity Grade BP/EP/USP
Physical Form Solid
Molecular Formula C6H12O6 · H2O
Linear Formula CH2OHCHO(CHOH)3CHOH·H2O
Formula Weight 198.17
MDL Number MFCD00149450
Synonym D-Glucose Monohydrate
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents