Please login to your online account to display your discounted pricing

Dextrose, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  191605

 View more versions of this product

Catalog No. 02002707

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Dextrose
Specific Gravity 1L = 1.54kg
Quantity 2.5kg
Name Note Anhydrous
CAS 50-99-7
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Powder
Molecular Formula C6H12O6
Formula Weight 180.16
Packaging Poly Bottle

Specifications

Chemical Name or Material Dextrose
CAS 50-99-7
Name Note Anhydrous
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C6H12O6
Formula Weight 180.16