Please login to your online account to display your discounted pricing

Dextrose, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$299.43 - $299.43

Chemical Identifiers

Chemical Name or Material Dextrose
CAS 50-99-7
Name Note Anhydrous
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C6H12O6
Formula Weight 180.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002707

 
avantor performance
191605
Poly Bottle 2.5kg Each for $299.43
Case of 4 Each for $1,197.72
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 180.16
Chemical Name or Material Dextrose
Physical Form Powder
CAS 50-99-7
Name Note Anhydrous
Packaging Poly Bottle
Quantity 2.5kg
Molecular Formula C6H12O6
Specific Gravity 1L = 1.54kg