missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dextrose Anhydrous (Crystalline Powder/EP/BP/USP), Fisher Chemical

$163.33 - $3,890.80

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00148912
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
D14-212 SDS View Documents Thermo Scientific
FLD14212
2.5kg Glass Bottle Each for $412.87
Case of 4 Each for $1,651.48
D14-50 SDS Accessories View Documents Thermo Scientific
FLD1450
50kg Fiber Drum Each for $3,890.80
D14-500 SDS View Documents Thermo Scientific
FLD14500
500g Glass Bottle Each for $163.33
Case of 6 Each for $979.98
Specifications

Chemical Identifiers

50-99-7
180.156
GZCGUPFRVQAUEE-SLPGGIOYSA-N
107526
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00148912
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
CHEBI:42758
C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Pass Test (BP/EP/USP)
Pass Test (BP/EP/USP)
Glass Bottle
52.5 to 53.3° (BP/EP), +52.6 to +53.2° (USP)
Negligible
White
2.5kg
50-99-7
CH2OH(CHOH)4CHO
2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
C(C(C(C(C(C=O)O)O)O)O)O
180.156
CHEBI:42758
Solid
Dextrose Anhydrous
Pass Test (EP/BP)
0.1% max. (USP)
Meets Requirements (BP/EP/USP)
0.1% max. (BP/EP)
1.0% Max. (BP/EP), 0.5% Max. (USP)
146.1°C
5.9
C6H12O6
MFCD00148912
GZCGUPFRVQAUEE-SLPGGIOYSA-N
(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
107526
180.16
BP/EP/USP
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0