Please login to your online account to display your discounted pricing

Dextrose Anhydrous (Crystalline Powder/EP/BP/USP), Fisher Chemical


Quantity 50kg
Packaging Fiber Drum

Manufacturer: thermo fisher scientific  FLD1450

 View more versions of this product

Catalog No. D14-50

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade BP/EP/USP
Melting Point 146.1°C
Formula Weight 180.16
Quantity 50kg
Physical Form Solid
Molecular Formula C6H12O6
pH 5.9
Water 1.0% Max. (BP/EP), 0.5% Max. (USP)
Color White
Specific Rotation +52.6 to +53.2° (USP)
Soluble Starch and Sulfites Pass Test (USP)
Identification Pass Test (BP)
Barium (Ba) Pass Test (BP)
Acidity Alkalinity Pass Test (BP)
Arsenic (As) 1ppm max. (BP)
Dextrins Pass Test (USP)
Sulfite 15ppm max. (BP)
Sulfate 0.025% max. (USP)
Pyrogens Pass Test (BP)
Residual Solvents Meets Requirements (BP)
Chloride 0.018% max. (USP)
Bacterial Endotoxins 1 EU/mg max. (EP)
Acidity Pass Test (USP)
Synonym Blood Sugar
Sulfated Ash 0.1% max. (BP)
Lead (Pb) 0.5ppm max. (BP)
Calcium (Ca) 200ppm max. (BP)
ChemAlert Storage Symbol Gray
MDL Number MFCD00148912
Chemical Name or Material Dextrose Anhydrous
CAS 50-99-7
Linear Formula CH2OH(CHOH)4CHO
Endotoxin 1 EU/mg max. (USP)
Heavy Metals (as Pb) 5ppm max. (USP)
Appearance of Solution Pass Test (BP)
Ignition Residue 0.1% max. (USP)
Additional Information Vapor Pressure: Negligible
Packaging Fiber Drum
Health Hazard 2 CAUTION!
Health Hazard 3 Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

Specifications

Chemical Name or Material Dextrose Anhydrous
CAS 50-99-7
Purity Grade BP/EP/USP
Physical Form Solid
Molecular Formula C6H12O6
Linear Formula CH2OH(CHOH)4CHO
Formula Weight 180.16
MDL Number MFCD00148912
Synonym Blood Sugar
Accessories

Accessories for Catalog Number: D1450

Item Mfr. No. Item Description Price Per Unit Quantity  
19-087809
Enpac™ Poly-Dolly™
spilkleen/cep
5300YE
Versatile all-in-one dolly, dispensing station and containment unit Each for $1,117.76
Accessories
 

Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

SDS & Safety Documents