Please login to your online account to display your discounted pricing

Dextrose, Anhydrous, ACS, EMD Millipore


Quantity 2.5kg
Packaging Plastic Bottle

Manufacturer: emd millipore corporation  DX01453

 View more versions of this product

Catalog No. MDX01453

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS/NF/USP
Formula Weight 180.16
Quantity 2.5kg
Packaging Plastic Bottle
Color White
Chemical Name or Material Dextrose, Anhydrous
Name Note For ACS analysis
Physical Form Powder
Molecular Formula CH2OH(CHOH)4CHO
Linear Formula CH2OH(CHOH)4CHO
CAS 50-99-7
Synonym D-Glucose

Specifications

Chemical Name or Material Dextrose, Anhydrous
Name Note For ACS analysis
CAS 50-99-7
Purity Grade ACS/NF/USP
Physical Form Powder
Molecular Formula CH2OH(CHOH)4CHO
Linear Formula CH2OH(CHOH)4CHO
Formula Weight 180.16
Synonym D-Glucose