missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Dextrin, precipitated by alcohol
Manufacturer: Alfa Aesar™ A1571736
Specifications
Dextrin | |
9004-53-9 | |
MFCD00081554 | |
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O | |
504.438 | |
500g |
precipitated by alcohol | |
C18H32O16 | |
14,2953 | |
FYGDTMLNYKFZSV-MRCIVHHJSA-N | |
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
62698 |
Chemical Identifiers
9004-53-9 | |
504.438 | |
FYGDTMLNYKFZSV-MRCIVHHJSA-N | |
62698 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O |
C18H32O16 | |
MFCD00081554 | |
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen | |
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
RTECSNumber : HH9450000
TSCA : TSCA