missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dextrin, precipitated by alcohol, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1571722
Description
Dextrin is used as adhesive in the manufacture of gummed tapes, textiles and paper. It is used in producing nutritional products. It is also used as a absorbents, binding agents and viscosity controlling agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dextrin | |
9004-53-9 | |
MFCD00081554 | |
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen | |
100 g | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O | |
504.438 | |
Hygroscopic |
precipitated by alcohol | |
C18H32O16 | |
14,2953 | |
Soluble in hot water (0.1g/ml). | |
FYGDTMLNYKFZSV-MRCIVHHJSA-N | |
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
62698 |
Chemical Identifiers
9004-53-9 | |
504.438 | |
FYGDTMLNYKFZSV-MRCIVHHJSA-N | |
62698 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O |
C18H32O16 | |
MFCD00081554 | |
dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen | |
(3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Safety and Handling
EINECSNumber : 232-675-4
RTECSNumber : HH9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only