Please login to your online account to display your discounted pricing

Dextrin, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  406285000

 View more versions of this product

Catalog No. AC406285000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Quantity 500g
Color White to pale yellow
Merck Index 15, 2955
EINECS Number 232-675-4
TSCA TSCA
RTECS Number HH9450000
Infrared Spectrum Authentic
MDL Number MFCD00081554
Chemical Name or Material Dextrin
Linear Formula (C6H10O5)x
CAS 9004-53-9
Packaging Plastic bottle
Sulfated Ash 0.5% max.

Specifications

Chemical Name or Material Dextrin
CAS 9004-53-9
Linear Formula (C6H10O5)x
MDL Number MFCD00081554