Learn More
Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000, Thermo Scientific™
Dextran sulfate sodium salt, C9H11NO2, CAS Number-9011-18-1, americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin, 25g, CHEBI:116735, BLFLLBZGZJTVJG-UHFFFAOYSA-N, ethyl 4-aminobenzoate, MFCD00081551, 165.192
Supplier: Thermo Fisher Scientific 441490050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Dextran sulfate sodium salt | |
9011-18-1 | |
MFCD00081551 | |
5g | |
CCOC(=O)C1=CC=C(C=C1)N | |
165.192 | |
CHEBI:116735 | |
Solubility in water: 100g/L (25°C). |
approx. M.W. 6, 500 - 10, 000 | |
C9H11NO2 | |
americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
ethyl 4-aminobenzoate | |
2337 | |
Glass bottle |
Chemical Identifiers
9011-18-1 | |
165.192 | |
BLFLLBZGZJTVJG-UHFFFAOYSA-N | |
2337 | |
ethyl 4-aminobenzoate |
C9H11NO2 | |
MFCD00081551 | |
americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin | |
CHEBI:116735 | |
CCOC(=O)C1=CC=C(C=C1)N |
RUO – Research Use Only