missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dextran sulfate sodium salt, approx. M.W. 6,500 - 10,000, Thermo Scientific™

Dextran sulfate sodium salt, C9H11NO2, CAS Number-9011-18-1, americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin, 25g, CHEBI:116735, BLFLLBZGZJTVJG-UHFFFAOYSA-N, ethyl 4-aminobenzoate, MFCD00081551, 165.192

Manufacturer:  Thermo Fisher Scientific AC441491000

 View more versions of this product

Catalog No. AC441491000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Dextran sulfate sodium salt
9011-18-1
MFCD00081551
100g
CCOC(=O)C1=CC=C(C=C1)N
165.192
CHEBI:116735
Solubility in water: 100g/L (25°C).
approx. M.W. 6, 500 - 10, 000
C9H11NO2
americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
Plastic bottle

Chemical Identifiers

9011-18-1
165.192
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
C9H11NO2
MFCD00081551
americaine, anaesthesin, anesthesin, benzocaine, ethoform, ethyl aminobenzoate, ethyl p-aminobenzoate, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(C=C1)N
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only