missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dextran sulfate sodium salt, approx. M.W. 500,000, ACROS Organics™

Manufacturer:  Acros Organics AC433241000

 View more versions of this product

Catalog No. AC433241000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Dextran sulfate sodium salt
C9H11NO2
approx. 50000g/mol
CCOC(=O)C1=CC=C(C=C1)N
165.192
CHEBI:116735
0.01% max.
0.1% max.
Glass bottle
40% to 50% ()
100g
9011-18-1
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
BLFLLBZGZJTVJG-UHFFFAOYSA-N
ethyl 4-aminobenzoate
2337
Powder
Authentic
10% max.
Solubility in water: 100g/L (25°C).
White to Yellow

Chemical Identifiers

9011-18-1
165.192
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
CHEBI:116735
CCOC(=O)C1=CC=C(C=C1)N
C9H11NO2
BLFLLBZGZJTVJG-UHFFFAOYSA-N
2337
ethyl 4-aminobenzoate
SDS Documents