Please login to your online account to display your discounted pricing

Dextran, MP Biomedicals™


Quantity 500g

Manufacturer: mp biomedicals  0210150890

 View more versions of this product

Catalog No. ICN10150890

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Dextran
CAS 9004-54-0
Physical Form White to slightly off white powder
Molecular Formula (C6H12O6)n
Formula Weight 35000 to 50000g/mol
MDL Number MFCD00130935
Synonym DSS
Recommended Storage Store at Room Temperature (15° to 30°C)

Specifications

Chemical Name or Material Dextran
CAS 9004-54-0
Physical Form White to slightly off white powder
Molecular Formula (C6H12O6)n
Formula Weight 35000 to 50000g/mol
MDL Number MFCD00130935
Synonym DSS
Accessories

Store at Room Temperature (15° to 30°C)