missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran 40 (Mw.=ca. 40,000), TCI America™
$121.16 - $1090.48
Chemical Identifiers
| CAS | 9004-54-0 |
|---|---|
| Molecular Formula | (C6H9O5)n(C6H10O5)m |
| Molecular Weight (g/mol) | 504.44 |
| MDL Number | MFCD00130935 |
| InChI Key | FZWBNHMXJMCXLU-UHFFFAOYNA-N |
| Synonym | dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal |
| PubChem CID | 4125253 |
| IUPAC Name | Dextran |
| SMILES | O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
Chemical Identifiers
| 9004-54-0 | |
| 504.44 | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| 4125253 | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O |
| (C6H9O5)n(C6H10O5)m | |
| MFCD00130935 | |
| dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal | |
| Dextran |
Specifications
| White | |
| 9004-54-0 | |
| MFCD00130935 | |
| 25 g | |
| O[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O | |
| 504.44 | |
| Dextran 40 (Mw.=ca. 40,000) |
| Crystal-Powder at 20°C | |
| (C6H9O5)n(C6H10O5)m | |
| dextran, dextran, mw-86.000 aver., 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy hexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-2-yl oxyhexanal, 2,3,4,5-tetrahydroxy-6-3,4,5-trihydroxy-6-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxymethyl tetrahydropyran-2-yl oxy-hexanal | |
| FZWBNHMXJMCXLU-UHFFFAOYNA-N | |
| Dextran | |
| 4125253 |
Safety and Handling
RTECSNumber : HH9246000
TSCA : Yes