Please login to your online account to display your discounted pricing

Dexrazoxane hydrochloride, Tocris™

Topoisomerase II inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material Dexrazoxane hydrochloride
Assay Percent Range >98%
Molecular Formula C11H16N4O4·HCl
Formula Weight 304.73
Synonym 4-[(2S)-2-(3, 5-Dioxopiperazin-1-yl)propyl]piperazine-2, 6-dione hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

385710

 
r & d systems
385710
10mg Each for $164.61

385750

 
r & d systems
385750
50mg Each for $682.67
Description & Specifications

Specifications

Formula Weight 304.73
Quantity 10mg
CAS 1263283-43-7
Chemical Name or Material Dexrazoxane hydrochloride
Synonym 4-[(2S)-2-(3, 5-Dioxopiperazin-1-yl)propyl]piperazine-2, 6-dione hydrochloride
Molecular Formula C11H16N4O4·HCl
Assay Percent Range >98%
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature