Thermo Scientific™ Dewar Flasks
Chemical resistant and withstand a wide temperature range.
Manufacturer:  Thermo Scientific™ 41501000EMD
 View more versions of this product
Catalog No. S34074A
Specifications Description & Specifications
Specifications
Polyethylene
HDPE
19.5cm
3.8 in.
33.81 oz., 1L
-321° to 212°F
7.7 in.
Dry solvent and hot baths
9.6cm
1L
9.9 in.
-196° to 100°C
33.81 oz.
25.14cm