Please login to your online account to display your discounted pricing

Devazepide, Tocris

Selective, orally active CCK1 receptor antagonist

$303.99 - $1,257.94

Chemical Identifiers

Chemical Name or Material Devazepide
Assay Percent Range >99%
Molecular Formula C25H20N4O2
Formula Weight 408.46
Synonym N-[(3S)-2, 3-Dihydro-1-methyl-2-oxo-5-phenyl-1H-1, 4-benzodiazepin-3-yl]-1H-indole-2-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

230410

 
r & d systems
230410
10mg Each for $303.99

230450

 
r & d systems
230450
50mg Each for $1,257.94
Description & Specifications

Specifications

Formula Weight 408.46
Quantity 10mg
Chemical Name or Material Devazepide
Assay Percent Range >99%
Molecular Formula C25H20N4O2
CAS 103420-77-5
Synonym N-[(3S)-2, 3-Dihydro-1-methyl-2-oxo-5-phenyl-1H-1, 4-benzodiazepin-3-yl]-1H-indole-2-carboxamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C