Please login to your online account to display your discounted pricing

Deuterium oxide, deuteration degree min. 99.96% for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$79.35 - $106.45

Chemical Identifiers

Chemical Name or Material Deuterium oxide
CAS 7789-20-0
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula D2O (Hill Formula)
Formula Weight 20.03
Synonym Heavy water
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1034280005

 
emd millipore corporation
1034280005
Glass ampoule 10 x 0.5mL Each for $79.35

M1034280009

 
emd millipore corporation
1034280009
Glass ampoule 10 x 0.75mL Each for $87.89

M1034280010

 
emd millipore corporation
1034280010
Septum ampoule 10mL Each for $106.45
Description & Specifications

Specifications

Formula Weight 20.03
Quantity 10 x 0.5mL
Color Colorless
Physical Form Liquid
Chemical Name or Material Deuterium oxide
Assay Percent Range 99.96% min.
Molecular Formula D2O (Hill Formula)
CAS 7789-20-0
Synonym Heavy water
Odor Odorless
Packaging Glass ampoule