Please login to your online account to display your discounted pricing

Deuterium oxide, deuteration degree min. 99.9% for NMR spectroscopy MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$46.41 - $1,183.57

Chemical Identifiers

Chemical Name or Material Deuterium oxide
CAS 7789-20-0
Physical Form Liquid
Assay Percent Range 99.90%
Molecular Formula D2O (Hill Formula)
Formula Weight 20.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1133660009

 
emd millipore corporation
1133660009
Glass bottle 10 x 0.75mL Each for $46.41

M1133660010

 
emd millipore corporation
1133660010
Septum ampoule 10mL Each for $48.79

M1133660025

 
emd millipore corporation
1133660025
Glass bottle 25mL Each for $89.04

M1133660500

 
emd millipore corporation
1133660500
Glass bottle 500mL Each for $1,183.57

M1133660100

 
emd millipore corporation
1133660100
Glass Bottle 100mL Each for $253.16
Description & Specifications

Specifications

Formula Weight 20.03
Quantity 10 x 0.75mL
Color Colorless
Physical Form Liquid
Chemical Name or Material Deuterium oxide
Assay Percent Range 99.90%
Molecular Formula D2O (Hill Formula)
CAS 7789-20-0
Odor Odorless
Packaging Glass bottle