Please login to your online account to display your discounted pricing

Deuterium oxide, 99.8% (Isotopic), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Deuterium Oxide
CAS 7789-20-0
Physical Form Liquid
Assay Percent Range 99.8% (Isotopic)
Molecular Formula D2O
Formula Weight 20.03
MDL Number MFCD00044636
Synonym Water, heavy
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1476418

  SDS
alfa aesar
1476418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1476430

  SDS
alfa aesar
1476430
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Formula Weight 20.03
Physical Form Liquid
Quantity 50g
EINECS Number 232-148-9
Density 1.105g/cm3 at 20°C
Odor Odorless
Assay Percent Range 99.8% (Isotopic)
Appearance Clear liquid
MDL Number MFCD00044636
CAS 7789-20-0
Molecular Formula D2O
Chemical Name or Material Deuterium oxide
Vapor Pressure 6 hPa at 25°C
Synonym Water, heavy