missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Deuterium oxide, 99.8% (Isotopic), Alfa Aesar™

Manufacturer:  Alfa Aesar 1476430

 View more versions of this product

Catalog No. AA1476430



Specifications

Specifications

Clear liquid
1.105g/cm3 at 20°C
6 hPa at 25°C
Colorless
250g
99.8% (Isotopic)
MFCD00044636
XLYOFNOQVPJJNP-ZSJDYOACSA-N
20.027
CHEBI:41981
Liquid
Deuterium Oxide
Odorless
101.4°C
3.8°C
7789-20-0
H2O
deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
O
24602
20.03

Chemical Identifiers

7789-20-0
20.027
XLYOFNOQVPJJNP-ZSJDYOACSA-N
24602
O
H2O
MFCD00044636
deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
CHEBI:41981
Safety and Handling

Safety and Handling

EINECSNumber : 232-148-9

SDS Documents