Please login to your online account to display your discounted pricing

Deuterium chloride 38% soln. in D2O, deuteration deg. min. 99.5%, for NMR, MagniSolv™, EMD Millipore™


Quantity 50mL
Packaging Glass bottle

Manufacturer: emd millipore corporation  8150180050

 View more versions of this product

Catalog No. M8150180050

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mL
Color Colorless
Physical Form Liquid
Molecular Formula DCl
Vapor Pressure 21.3 hPa (20°C)
Chemical Name or Material Deuterium chloride
Name Note 20% soln. in D2O
Odor Stinging
Packaging Glass bottle
Assay Percent Range 99.5% min.

Specifications

Chemical Name or Material Deuterium chloride
Name Note 20% soln. in D2O
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula DCl