Please login to your online account to display your discounted pricing

Deuterium chloride 38% soln. in D2O, deuteration deg. min. 99.5%, for NMR, MagniSolv™, EMD Millipore™

$235.87 - $773.92

Chemical Identifiers

Chemical Name or Material Deuterium chloride
Name Note 20% soln. in D2O
Physical Form Liquid
Assay Percent Range 99.5% min.
Molecular Formula DCl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M8150180010

 
emd millipore corporation
8150180010
Glass ampoule 10mL Each for $235.87

M8150180050

 
emd millipore corporation
8150180050
Glass bottle 50mL Each for $773.92
Description & Specifications

Specifications

Chemical Name or Material Deuterium chloride
Name Note 20% soln. in D2O
Odor Stinging
Packaging Glass ampoule
Vapor Pressure 21.3 hPa (20°C)
Physical Form Liquid
Quantity 10mL
Assay Percent Range 99.5% min.
Color Colorless
Molecular Formula DCl