Please login to your online account to display your discounted pricing

Determining Density Lab


Archimedes' Principle and density activities.

Manufacturer: fisher science education  AZ-20-3061

 View more versions of this product

Catalog No. S66689

  • / Each
 

Description & Specifications

Specifications

Includes Comprehensive student and teacher guides containing a variety of demonstrations and activities
Quantity 1 kit
Product Type Determining density kit
For Use With (Application) To determine the volume and density of objects, calculate specific gravity
Grade 7 to10