missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Determining Density Lab

Archimedes' Principle and density activities.

$66.85 - $66.85

Specifications

For Use With (Application) To determine the volume and density of objects, calculate specific gravity
Product Type Determining density kit
Grade 7 to10
Includes Comprehensive student and teacher guides containing a variety of demonstrations and activities
View More Specs

Includes: Comprehensive teacher and student guides with demonstrations and activities


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S66689 Fisher Science Education
AZ-20-3061
1 kit Each for $66.85
Specifications

Description

  • Determine the volume and density of an object with the displacement of liquid method
  • Calculate specific gravity and prove Archimedes' Principle

Description & Specifications

Specifications

To determine the volume and density of objects, calculate specific gravity
Determining density kit
7 to10
Comprehensive student and teacher guides containing a variety of demonstrations and activities