Please login to your online account to display your discounted pricing

Alfa Aesar™ Desyl chloride, 98%

Chemical Identifiers

Chemical Name or Material Desyl chloride
CAS 447-31-4
Molecular Formula C14H11ClO
Formula Weight 230.69
MDL Number MFCD00000858
UN Number UN3261
Beilstein 744474
Synonym alpha-Chlorodeoxybenzoin; 2-Chloro-2-phenylacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2177806

  SDS
alfa aesar™
B2177806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2177814

  SDS
alfa aesar™
B2177814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2177822

  SDS
alfa aesar™
B2177822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.69
Chemical Name or Material Desyl chloride
TSCA TSCA
Assay 98%
Molecular Formula C14H11ClO
MDL Number MFCD00000858
CAS 447-31-4
UN Number UN3261
Beilstein 744474
Synonym alpha-Chlorodeoxybenzoin; 2-Chloro-2-phenylacetophenone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 64° to 69°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
RTECS Number AM6825000
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.