missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dess-Martin periodinane, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1577906

 View more versions of this product

Catalog No. AAL1577906


Description

Description

Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.

Solubility
Soluble in chloroform, acetone, acetonitrile and methylene chloride. Slightly soluble in ether and hexane.

Notes
Store in cool place. Air and moisture sensitive. Incompatible with strong reducing agents, combustible material and water.
Specifications

Specifications

Dess-Martin periodinane
130°C to 132°C (decomposition)
MFCD00130127
UN1479
Air, light, and moisture sensitive
dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane
NKLCNNUWBJBICK-UHFFFAOYSA-N
1,1-bis(acetyloxy)-3-oxo-3H-1λ⁵,2-benziodaoxol-1-yl acetate
159087
≥95%
87413-09-0
C13H13IO8
5 g
4548207
14,2933
Soluble in chloroform,acetone,acetonitrile and methylene chloride. Slightly soluble in ether and hexane.
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
424.14
424.15

Chemical Identifiers

87413-09-0
424.14
NKLCNNUWBJBICK-UHFFFAOYSA-N
CC(=O)O[I]1(OC(C)=O)(OC(C)=O)OC(=O)C2=CC=CC=C12
C13H13IO8
MFCD00130127
159087
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H272-H315-H319-H335

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only