missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Desoxybenzoin p-toluenesulfonylhydrazone, 97%

Manufacturer:  Alfa Aesar™ H5344714

 View more versions of this product

Catalog No. AAH5344714



Description

Description

Applications
Desoxybenzoin p-toluenesulfonylhydrazone is used as the reagent.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Desoxybenzoin p-toluenesulfonylhydrazone
25g
C21H20N2O2S
desoxybenzoin p-toluenesulfonylhydrazone
CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC2=CC=CC=C2)C3=CC=CC=C3
364.463
364.46
139°C to 143°C
19816-85-4
MFCD06411289
INMYHVHTZKDKDK-DQRAZIAOSA-N
N-[(Z)-1,2-diphenylethylideneamino]-4-methylbenzenesulfonamide
5834443
97%

Chemical Identifiers

19816-85-4
364.463
INMYHVHTZKDKDK-DQRAZIAOSA-N
5834443
CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC2=CC=CC=C2)C3=CC=CC=C3
C21H20N2O2S
MFCD06411289
desoxybenzoin p-toluenesulfonylhydrazone
N-[(Z)-1,2-diphenylethylideneamino]-4-methylbenzenesulfonamide
Documents