missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Desoxybenzoin p-toluenesulfonylhydrazone, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5344706
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsDesoxybenzoin p-toluenesulfonylhydrazone is used as the reagent.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Desoxybenzoin p-toluenesulfonylhydrazone | |
139°C to 143°C | |
MFCD06411289 | |
desoxybenzoin p-toluenesulfonylhydrazone, n-e-1,2-diphenylethylideneamino-4-methylbenzenesulfonamide, toluene-4-sulfonic acid dibenzyl-alpha-ylidenehydrazone, n'-1z-1,2-diphenylethylidene-4-methylbenzenesulfonohydrazide | |
INMYHVHTZKDKDK-DQRAZIAOSA-N | |
N-[(Z)-1,2-diphenylethylideneamino]-4-methylbenzenesulfonamide | |
5834443 | |
97% |
19816-85-4 | |
C21H20N2O2S | |
5 g | |
Soluble in water. | |
CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC2=CC=CC=C2)C3=CC=CC=C3 | |
364.463 | |
364.46 |
Chemical Identifiers
19816-85-4 | |
364.463 | |
INMYHVHTZKDKDK-DQRAZIAOSA-N | |
5834443 | |
CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC2=CC=CC=C2)C3=CC=CC=C3 |
C21H20N2O2S | |
MFCD06411289 | |
desoxybenzoin p-toluenesulfonylhydrazone, n-e-1,2-diphenylethylideneamino-4-methylbenzenesulfonamide, toluene-4-sulfonic acid dibenzyl-alpha-ylidenehydrazone, n'-1z-1,2-diphenylethylidene-4-methylbenzenesulfonohydrazide | |
N-[(Z)-1,2-diphenylethylideneamino]-4-methylbenzenesulfonamide |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures