Please login to your online account to display your discounted pricing

Desformylflustrabromine hydrochloride, Tocris

Positive allosteric modulator of α4β2

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Desformylflustrabromine hydrochloride
Assay Percent Range >98%
Molecular Formula C16H21BrN2·HCl
Formula Weight 357.72
Synonym 6-Bromo-2-(1, 1-dimethyl-2-propenyl)-N-1H-indole-3-ethanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

332850

 
r & d systems
332850
50mg Each for $915.10

332810

 
r & d systems
332810
10mg Each for $218.74
Description & Specifications

Specifications

Formula Weight 357.72
Quantity 50mg
Chemical Name or Material Desformylflustrabromine hydrochloride
Assay Percent Range >98%
Molecular Formula C16H21BrN2·HCl
CAS 951322-11-5
Synonym 6-Bromo-2-(1, 1-dimethyl-2-propenyl)-N-1H-indole-3-ethanamine hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C