Please login to your online account to display your discounted pricing

Alfa Aesar™ [Des-His&superscript_1;, Glu&superscript_9;] Glucagon

For cellular and molecular biology applications

Specifications

Product Type [Des-His1, Glu9] Glucagon
Molecular Weight 3359.60
Storage Requirements -30° to -10°C
Sequence Ser-Gln-Gly-Thr-Phe-Thr-Ser-Glu-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66456EXD

 
alfa aesar™
J66456EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66456MCR

 
alfa aesar™
J66456MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Des-His1, Glu9] Glucagon
Molecular Weight 3359.60
Storage Requirements -30° to -10°C
Sequence Ser-Gln-Gly-Thr-Phe-Thr-Ser-Glu-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr
Format Solid
Regulatory Status RUO
Purification Method HPLC