Please login to your online account to display your discounted pricing

Alfa Aesar™ Dermenkephalin

For cellular and molecular biology applications

Specifications

Product Type Dermenkephalin
Molecular Weight 955.15
Storage Requirements -30° to -10°C
Sequence Tyr-D-Met-Phe-His-Leu-Met-Asp-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66523LB0

 
alfa aesar™
J66523LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66523MCR

 
alfa aesar™
J66523MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Dermenkephalin
Molecular Weight 955.15
Storage Requirements -30° to -10°C
Sequence Tyr-D-Met-Phe-His-Leu-Met-Asp-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC