Please login to your online account to display your discounted pricing

Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP25145

 View more versions of this product

Catalog No. BP25145

  • / Each
  • / Case of 9 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 6000000
Quantity 5g
Physical Form Solid
Color Off-white
Protein Pass Test
Polymerization Pass Test
Recommended Storage 4°C
Name Note From Salmon Sperm
Loss on Drying 14% max.
Packaging Amber Glass
Identification Pass Test
Phosphorus Approx. 8 to 9%
Chemical Name or Material Deoxyribonucleic Acid Sodium Salt
Nitrogen (N) Approx. 14%
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Deoxyribonucleic Acid Sodium Salt
Name Note From Salmon Sperm
Physical Form Solid
Formula Weight 6000000
Accessories
 

4°C

SDS & Safety Documents