Please login to your online account to display your discounted pricing

Deoxyribonucleic Acid Sodium Salt, Fisher BioReagents


Quantity 250mg
Packaging Amber Glass

Manufacturer: fisher scientific company  BP2514250

 View more versions of this product

Catalog No. BP2514250

  • / Each
  • / Case of 10 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 6000000
Quantity 250mg
Physical Form Solid
Color Off-white
Chemical Name or Material Deoxyribonucleic Acid Sodium Salt
Name Note From Salmon Sperm
Identification Pass Test
ChemAlert Storage Symbol Gray
Loss on Drying 14% max.
Nitrogen (N) Approx. 14%
Packaging Amber Glass
Phosphorus Approx. 8 to 9%
Polymerization Pass Test
Protein Pass Test
Recommended Storage 4°C

Specifications

Chemical Name or Material Deoxyribonucleic Acid Sodium Salt
Name Note From Salmon Sperm
Physical Form Solid
Formula Weight 6000000
Accessories
 

4°C

SDS & Safety Documents