missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Deoxyribonucleic acid sodium salt, calf thymus

Suitable for use as a substrate in DNase assays

$106.09 - $565.96

Chemical Identifiers

CAS 91080-16-9
MDL Number MFCD00151698
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ64400MC View Documents Alfa Aesar™
J64400MC
100mg Each for $106.09
 
AAJ6440003 View Documents Alfa Aesar™
J6440003
1g Each for $565.96
 
Description

Description

Applications
Deoxyribonucleic acid sodium salt, is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. Calf thymus DNA is exceptionally useful as a substrate for DNA polymerase assays, in the amplification of very long fragments and as a carrier DNA for precipitations. It was used as a standard to determine DNA content in gels and for DNA quantification by fluorescent assay.

Solubility
Soluble in water.

Notes
Highly polymerized. Hyperchromicity is at least 27%.
Specifications

Chemical Identifiers

91080-16-9
MFCD00151698

Specifications

White
91080-16-9
Lyophilized Powder
100mg
MFCD00151698
Deoxyribonucleic acid sodium salt, calf thymus
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C