Please login to your online account to display your discounted pricing

American Educational Products Density Kit


Learn measurement, density, and the metric system

Manufacturer: american educational products  HUB3132

 View more versions of this product

Catalog No. S53006A

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Grade 6 to 12
Product Type Density kit
For Use With (Application) To learn measurement, the metric system, how to use the formula: Density=Mass/Volume
Includes 3 x 1 x 0.25 in. Aluminum bar, two 0.5 in. Aluminum cubes, Steel sphere, Glass sphere, 15cm scale